Glycin (H-Gly-OH) 99

Lieferant: Apollo Scientific
Achtung

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

BIG0709-1KG BIG0709-5KG
APOSBIG0709-1KGEA 78.4 CHF
APOSBIG0709-1KG APOSBIG0709-5KG
Glycin (H-Gly-OH) 99
Glycin (H-Gly-OH)

Component of tris-glycine-sds running buffers forpolyacrylamide gel electrophoresis.


  • Ph. Eur.
  • Pyrogen free

Formel: NH₂CH₂COOH
MW: 75.07 g/mol
Siedepunkt: 181
Melting Pt: 240…245 °C (Zers.)
Dichte: 1.595 g/cm³ (25 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00008131
CAS-Nummer: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR