G418 Disulfat

Lieferant: Apollo Scientific
Gefahr

Synonyms: G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate

BIG0175-1G BIG0175-5G
APOSBIG0175-1GEA 38.3 CHF
APOSBIG0175-1G APOSBIG0175-5G
G418 Disulfat
G418 disulphate

Aminoglycoside antibiotic.

Formel: C₂₀H₄₄N₄O₁₈S₂
Lager Temperatur: Kühlschrank
MDL Number: MFCD00058314
CAS-Nummer: 108321-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR