D(+)-Glucose, anhydrous

Lieferant: Apollo Scientific
Achtung

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

BIFS0119-1KG BIFS0119-2.5KG BIFS0119-500G
APOSBIFS0119-1KGEA 59.2 CHF
APOSBIFS0119-1KG APOSBIFS0119-2.5KG APOSBIFS0119-500G
D(+)-Glucose, anhydrous
D(+)-Glucose
Formel: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 153…156 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00148912
CAS-Nummer: 50-99-7
EINECS: 200-075-1
Merck Index: 14,04459

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR