L(+)-Arabinose 98%

Lieferant: Apollo Scientific
Achtung

Synonyms: Pectinose, Pectin Sugar

BIFS0102-100G BIFS0102-1KG BIFS0102-250G
APOSBIFS0102-100GEA 78.4 CHF
APOSBIFS0102-100G APOSBIFS0102-1KG APOSBIFS0102-250G
L(+)-Arabinose 98%
L(+)-Arabinose
Formel: C₅H₁₀O₅
MW: 150.13 g/mol
Siedepunkt: 331
Melting Pt: 160…163 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00067709
CAS-Nummer: 5328-37-0
EINECS: 201-767-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR