N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimid Hydrochlorid

Lieferant: Apollo Scientific
Gefahr

Synonyms: N'-(Ethylcarbonimidoyl)-N,N-dimethylpropane-1,3-diamine monohydrochloride, N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide monohydrochloride, 1-Ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochloride, EDC hydrochloride, EDC, EDAC HCl, EDAC hydrochloride, 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, WSC HCl, EDC HCl

BIE1308-100G BIE1308-500G BIE1308-1KG
APOSBIE1308-100GEA 31.4 CHF
APOSBIE1308-100G APOSBIE1308-500G APOSBIE1308-1KG
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimid Hydrochlorid
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimid Hydrochlorid

EDAC is a water soluble condensing reagent.

Formel: C₈H₁₇N₃·HCl
MW: 191.7 g/mol
Lager Temperatur: Gefrierschrank
MDL Number: MFCD00012503
CAS-Nummer: 25952-53-8
EINECS: 247-361-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR