1,2,3,4-Tetra-O-acetyl-ɑ-L-fucopyranose 98% min

Lieferant: Apollo Scientific

Synonyms: 1,2,3,4-Tetra-O-acetyl-α-L(−)-fucose, Tetra-O-acetyl-α-L(−)fucopyranose, ɑ-L-Fucopyranose 1,2,3,4-Tetraacetate

BICL4254-1G BICL4254-2G BICL4254-5G
APOSBICL4254-1GEA 337 CHF
APOSBICL4254-1G APOSBICL4254-2G APOSBICL4254-5G
1,2,3,4-Tetra-O-acetyl-ɑ-L-fucopyranose 98% min
1,2,3,4-Tetra-O-acetyl-alpha-L-fucopyranose
Formel: C₁₄H₂₀O₉
MW: 332.31 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00069791
CAS-Nummer: 64913-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR