Methyl 1,2,3,4-tetra-O-pivaloyl-β-D-glucopyranosyluronate 98

Lieferant: Apollo Scientific

Synonyms: 1,2,3,4-Tetra-O-pivaloyl-β-D-glucuronic acid methyl ester

BICL4142-1G BICL4142-250MG BICL4142-500MG
APOSBICL4142-1GEA 1520 CHF
APOSBICL4142-1G APOSBICL4142-250MG APOSBICL4142-500MG
Methyl 1,2,3,4-tetra-O-pivaloyl-β-D-glucopyranosyluronate 98
Methyl 1,2,3,4-tetra-O-pivaloyl-β-D-glucopyranosyluronate
Formel: C₂₇H₄₄O₁₁
MW: 544.632 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD15145134
CAS-Nummer: 86448-91-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR