Chloramphenicol 99

Lieferant: Apollo Scientific
Gefahr

Synonyms: Chloromycetin®, Tega-Cetin, (R,R)-Chloramphenicol

BIC0113-100G BIC0113-250G
APOSBIC0113-100GEA 227 CHF
APOSBIC0113-100G APOSBIC0113-250G
Chloramphenicol 99
Chloramphenicol

Bacteriostatic antibiotic with activity against gram-positive and gram-negative bacteria.


  • Cell culture tested

Formel: Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
MW: 323.13 g/mol
Siedepunkt: 645
Melting Pt: 148…152 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00078159
CAS-Nummer: 56-75-7
EINECS: 200-287-4
Merck Index: 13,02087

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR