Anti-Fyn Rabbit Monoclonal Antibody [clone: ARC1459]

Lieferant: ANTIBODIES.COM
A307345-100
ANTIA307345-100EA 525 CHF
ANTIA307345-100
Anti-Fyn Rabbit Monoclonal Antibody [clone: ARC1459]
Antikörper
Rabbit monoclonal [ARC1459] antibody to Fyn for WB with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:2000

Type: Primary
Antigen: Fyn
Clonality: Monoclonal
Clone: ARC1459
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR