Anti-Fyn Rabbit Polyclonal Antibody

Lieferant: ANTIBODIES.COM
A10990-100
ANTIA10990-100EA 535 CHF
ANTIA10990-100
Anti-Fyn Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody to Fyn (phospho Ser21) for WB with samples derived from human.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:2000

Type: Primary
Antigen: Fyn
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

TECHNISCHE DATEN

Antigen-Symbol Fyn
Environmentally Preferable
Antigen-Name C syn protooncogene
Antigen-Synonyme FYN_HUMAN|Protein tyrosine kinase fyn|p59-Fyn|Proto-oncogene c-Fyn|Fyn|FYN oncogene related to SRC FGR YES|OKT3 induced calcium influx regulator|Proto oncogene tyrosine protein kinase fyn|P59 FYN
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Unconjugated
Reaktivität Human
Wirt Rabbit
Gen-ID UniprotID# P06241
Isotyp IgG
Western blot Yes
Modification phospho Ser21
Immunogen A synthetic phosphorylated peptide around S21 of human Phospho-Fyn-S21 (NP_002028.1)
Sequence DGSLN
Form Liquid
Molecular weight 60 kDa
Reinigung Affinity purification
Lagerungspuffer Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,02% Sodium Azide
Lagertemperatur Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
Transporttemperatur Shipped on blue ice at 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR