Surgical gloves, polyisoprene, sterile, Gammex® Non-Latex PI Micro
Lieferant: ANSELL MEDICAL PRODUCTS
340057055 340057060 340057065 340057070 340057075 340057080 340057085 340057090
ANSM340057055EA 220 CHF
ANSM340057055 ANSM340057060 ANSM340057065 ANSM340057070 ANSM340057075 ANSM340057080 ANSM340057085 ANSM340057090
Surgical gloves, polyisoprene, sterile, Gammex® Non-Latex PI Micro
Handschuhe
These thin gloves made from synthetic polyisoprene offer an optimal level of sensitivity, comfort, and protection against natural rubber latex allergies.
  • Powder-free, micro-textured finish
  • DPG-free polyisoprene option for healthcare workers suffering from latex allergies and chemical allergies
  • White colour makes it ideal as a double-gloving solution, together with the green GAMMEX® Non-Latex PI underglove
  • Beaded cuffs with adhesive band
  • External surface: Micro-textured and chlorinated and siliconised; internal surface: PU coated and siliconised