Cooking thermometer
Lieferant: Alla France
801.060-GB
ALLA801.060-GBEA 54.1 CHF
ALLA801.060-GB
Cooking thermometer
Thermometer Analoge Thermometer
Internal scale glass thermometer: −10 to +60 °C , for baking, milk and wine.
  • White polypropylene alimentary protection sleeve