Brilliant II SYBR® Master mix
Lieferant: AGILENT
600825 600826 600828 600831 600834
AGLS600825EA 1258 CHF
AGLS600825 AGLS600826 AGLS600828 AGLS600831 AGLS600834
Brilliant II SYBR® Master mix
Nukleinsäure-Reagenzien qPCR/RT-qPCR Enzyme und Kits
Brilliant II SYBR® QPCR and QRT-PCR reagents were developed for improved sensitivity of detection, ensuring reproducible quantification even at low target concentrations.
  • Earlier Ct detection
  • Improved data reproducibility
  • Greater flexibility for use with numerous different templates and targets
  • More accurate quantification results
  • Developed on the Agilent Mx QPCR systems
When compared to other commercially available QPCR reagents, we observe earlier threshold cycle (Ct) detection at high and low template concentrations as well as improved reaction efficiency and replicate reproducibility across the entire range of template concentrations.
Bitte beachten For research use only. Not for use in diagnostic procedures.
Order Now
TECHNISCHE DATEN
Environmentally Preferable
Anwendung RT-qPCR dye qPCR dye RT-qPCR dye
One step Ja