Magnesiumchlorid 0.025 mol/l (0.05 N) aqueous solution

Lieferant: Thermo Fisher Scientific

AB-0359
ADVRAB-0359EA 84.9 CHF
ADVRAB-0359
Magnesiumchlorid 0.025 mol/l (0.05 N) aqueous solution
Magnesium chloride
Formel: MgCl₂
MW: 95.21 g/mol
Siedepunkt: 1412
Melting Pt: 715
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011106
CAS-Nummer: 7786-30-3
EINECS: 232-094-6
Merck Index: 13,05684

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR