2-Chlor-5-(chlormethyl)pyridin

Lieferant: ACTIVATE SCIENTIFIC

AS7018G100
ACTIAS7018G100EA 0 CHF
ACTIAS7018G100
2-Chlor-5-(chlormethyl)pyridin
2-Chloro-5-(chloromethyl)pyridine
Formel: C₆H₅Cl₂N
MW: 162,02 g/mol
Melting Pt: 37 °C
MDL Number: MFCD00125366
CAS-Nummer: 70258-18-3
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR