4-Chlorpicolinsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Chloropyridine-2-carboxylic acid

AS6004G25
ACTIAS6004G25EA 39 CHF
ACTIAS6004G25
4-Chlorpicolinsäure
4-Chlorpicolinsäure
Formel: C₆H₄ClNO₂
MW: 157,56 g/mol
MDL Number: MFCD00191400
CAS-Nummer: 5470-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR