4-(Tributylstannyl)-3,6-dihydro-2H-pyran

Lieferant: ACTIVATE SCIENTIFIC
Gefahr

AS51876G1 AS51876G5
ACTIAS51876G1EA 402 CHF
ACTIAS51876G1 ACTIAS51876G5
4-(Tributylstannyl)-3,6-dihydro-2H-pyran
4-(Tributylstannyl)-3,6-dihydro-2H-pyran
Formel: C₁₇H₃₄OSn
MDL Number: MFCD18325166
CAS-Nummer: 535924-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR