2-Chlor-4-methylpyrimidin

Lieferant: ACTIVATE SCIENTIFIC

AS5149G5 AS5149G100
ACTIAS5149G5EA 0 CHF
ACTIAS5149G5 ACTIAS5149G100
2-Chlor-4-methylpyrimidin
2-Chloro-4-methylpyrimidine
Formel: C₅H₅ClN₂
MDL Number: MFCD00054434
CAS-Nummer: 13036-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR