4-Brom-2,5-dimethoxybenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS51284G5
ACTIAS51284G5EA 0 CHF
ACTIAS51284G5
4-Brom-2,5-dimethoxybenzaldehyd
4-Bromo-2,5-dimethoxybenzaldehyde
Formel: C₉H₉BrO₃
MW: 245.08 g/mol
MDL Number: MFCD00156964
CAS-Nummer: 31558-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR