2-Brom-3-fluorpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS50690G1 AS50690G5
ACTIAS50690G1EA 0 CHF
ACTIAS50690G1 ACTIAS50690G5
2-Brom-3-fluorpyridin
2-Bromo-3-fluoropyridine
Formel: C₅H₃BrFN
MW: 175.99 g/mol
MDL Number: MFCD04114128
CAS-Nummer: 40273-45-8
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR