1-Boc-4-iodpiperidin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-BOC-4-iodo-piperidine, tert-Butyl 4-iodopiperidine-1-carboxylate

AS5035G1 AS5035G25 AS5035G5
ACTIAS5035G1EA 0 CHF
ACTIAS5035G1 ACTIAS5035G25 ACTIAS5035G5
1-Boc-4-iodpiperidin
1-Boc-4-iodpiperidin
Formel: C₁₀H₁₈INO₂
MW: 311.16 g/mol
MDL Number: MFCD04115041
CAS-Nummer: 301673-14-3

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR