3-(Boc-amino)phenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Aminobenzeneboronic acid, N-BOC protected, 3-(N-Boc-amino)phenylboronic acid, (3-((tert-Butoxycarbonyl)amino)phenyl)boronic acid, 3-Boronoaniline, N-BOC protected, 3-[(tert-Butoxycarbonyl)amino]benzeneboronic acid, 3-(Boc-amino)benzeneboronic acid

AS50233G1 AS50233G5
ACTIAS50233G1EA 0 CHF
ACTIAS50233G1 ACTIAS50233G5
3-(Boc-amino)phenylboronsäure
3-(Boc-amino)phenylboronsäure
Formel: C₁₁H₁₆BNO₄
MW: 237.06 g/mol
MDL Number: MFCD03411945
CAS-Nummer: 380430-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR