2-Fluor-3-formylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Fluoronicotinaldehyde

AS50174G1 AS50174G5
ACTIAS50174G1EA 0 CHF
ACTIAS50174G1 ACTIAS50174G5
2-Fluor-3-formylpyridin
2-Fluor-3-formylpyridin
Formel: C₆H₄FNO
MW: 125.1 g/mol
MDL Number: MFCD03095270
CAS-Nummer: 36404-90-7

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR