4-Iod-1-methylimidazol-5-carbaldehyd

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Iodo-1-methyl-1H-imidazole-5-carbaldehyde

AS50086G1
ACTIAS50086G1EA 0 CHF
ACTIAS50086G1
4-Iod-1-methylimidazol-5-carbaldehyd
4-Iod-1-methylimidazol-5-carbaldehyd
Formel: C₅H₅IN₂O
MW: 236.0105 g/mol
MDL Number: MFCD01632219
CAS-Nummer: 189014-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR