3-Brom-2-nitroanilin

Lieferant: ACTIVATE SCIENTIFIC

AS40316G5
ACTIAS40316G5EA 0 CHF
ACTIAS40316G5
3-Brom-2-nitroanilin
3-Brom-2-nitroanilin
Formel: C₆H₅BrN₂O₂
MW: 217.02 g/mol
MDL Number: MFCD08063248
CAS-Nummer: 7138-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR