3-Brom-5-(trifluormethyl)anilin

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: 3-Amino-5-bromo-α,α,α-trifluorotoluene, 5-Bromo-α,α,α-trifluoro-m-toluidine, 3-Amino-5-bromobenzotrifluoride

AS40306G100
ACTIAS40306G100EA 0 CHF
ACTIAS40306G100
3-Brom-5-(trifluormethyl)anilin
3-Brom-5-(trifluormethyl)anilin
Formel: C₇H₅BrF₃N
MW: 240.02 g/mol
MDL Number: MFCD00236205
CAS-Nummer: 54962-75-3
EINECS: 259-412-6
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR