3-(Aminomethyl)pyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Pyridylmethylamine, (3-Pyridyl)methylamine, 3-Picolamine, Pyridin-3-ylmethanamine, 3-Picolylamine

AS40267G100
ACTIAS40267G100EA 0 CHF
ACTIAS40267G100
3-(Aminomethyl)pyridin
3-(Aminomethyl)pyridine
Formel: C₆H₈N₂
MW: 108,14 g/mol
Siedepunkt: 226 °C (1013 hPa)
Melting Pt: –21 °C
Dichte: 1,0712 g/cm³ (20 °C)
MDL Number: MFCD00006412
CAS-Nummer: 3731-52-0
EINECS: 223-091-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR