α,α,α,α',α',α'-Hexafluor-2,5-xylidin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,5-Bis(trifluoromethyl)aniline

AS40122G100
ACTIAS40122G100EA 0 CHF
ACTIAS40122G100
α,α,α,α',α',α'-Hexafluor-2,5-xylidin
α,α,α,α',α',α'-Hexafluor-2,5-xylidin
Formel: C₈H₅F₆N
MW: 229.12 g/mol
Siedepunkt: 173…177 °C (1013 hPa)
Dichte: 1.479 g/cm³ (20 °C)
MDL Number: MFCD00074940
CAS-Nummer: 328-93-8
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR