Methyl-8-hydroxy-1-naphthoat

Lieferant: ACTIVATE SCIENTIFIC

AS36835G1
ACTIAS36835G1EA 0 CHF
ACTIAS36835G1
Methyl-8-hydroxy-1-naphthoat
Methyl-8-hydroxy-1-naphthoat
Formel: C₁₂H₁₀O₃
MW: 202,21 g/mol
MDL Number: MFCD00665832
CAS-Nummer: 5247-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR