1-Benzyloxy-3-methyl-2-nitrobenzol

Lieferant: ACTIVATE SCIENTIFIC

AS35192G5
ACTIAS35192G5EA 0 CHF
ACTIAS35192G5
1-Benzyloxy-3-methyl-2-nitrobenzol
1-Benzyloxy-3-methyl-2-nitrobenzol
Formel: C₁₄H₁₃NO₃
MW: 243.2579 g/mol
MDL Number: MFCD00272556
CAS-Nummer: 61535-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR