4-(Ethoxycarbonylmethyl)phenylboronsäurepinakolester

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Ethyl 2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate, 2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetic acid ethyl ester, Ethyl 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetate, 2-[(4-Ethoxycarbonylmethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-(Ethoxycarbonylmethyl)phenylboronic acid Pinacol ester

AS33906G1 AS33906G5
ACTIAS33906G1EA 0 CHF
ACTIAS33906G1 ACTIAS33906G5
4-(Ethoxycarbonylmethyl)phenylboronsäurepinakolester
4-(Ethoxycarbonylmethyl)phenylboronsäurepinakolester
Formel: C₁₆H₂₃BO₄
MW: 290.1624 g/mol
MDL Number: MFCD08704684
CAS-Nummer: 859169-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR