3-Aminoisoxazol

Lieferant: ACTIVATE SCIENTIFIC

AS33411G1 AS33411G5
ACTIAS33411G1EA 0 CHF
ACTIAS33411G1 ACTIAS33411G5
3-Aminoisoxazol
3-Aminoisoxazole
Formel: C₃H₄N₂O
MDL Number: MFCD00038814
CAS-Nummer: 1750-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR