L(+)-Arabinose

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Pectinose, Pectin Sugar

AS33109G100
ACTIAS33109G100EA 0 CHF
ACTIAS33109G100
L(+)-Arabinose
L(+)-Arabinose
Formel: C₅H₁₀O₅
Siedepunkt: 331 °C (1013 hPa)
Melting Pt: 158…160 °C
Dichte: 1.585 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00067709
CAS-Nummer: 5328-37-0
EINECS: 226-214-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ad925ac21d4f12371621b1c44dbb6cda0a12da8&vid=6b9738c515009e5720388ea307490836c3b8afa8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR