3-Methoxy-2-nitropyridin

Lieferant: ACTIVATE SCIENTIFIC

AS33073G10
ACTIAS33073G10EA 0 CHF
ACTIAS33073G10
3-Methoxy-2-nitropyridin
3-Methoxy-2-nitropyridine
Formel: C₆H₆N₂O₃
MW: 154,13 g/mol
MDL Number: MFCD00222276
CAS-Nummer: 20265-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR