D(-)-Ribose

Lieferant: ACTIVATE SCIENTIFIC

AS32990G100
ACTIAS32990G100EA 0 CHF
ACTIAS32990G100
D(-)-Ribose
D(-)-Ribose
Formel: C₅H₁₀O₅
Siedepunkt: 331 °C (1013 hPa)
Melting Pt: 90…95 °C
Dichte: 0.8 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00135453
CAS-Nummer: 50-69-1
EINECS: 200-059-4
Merck Index: 12,08371

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR