Hydroxyterephthalsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Hydroxyterephthalic acid

AS32843G1 AS32843G5
ACTIAS32843G1EA 0 CHF
ACTIAS32843G1 ACTIAS32843G5
Hydroxyterephthalsäure
Hydroxyterephthalsäure
Formel: C₈H₆O₅
MW: 182.1302 g/mol
MDL Number: MFCD09835368
CAS-Nummer: 636-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR