4-Chlor-2-fluorphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Chloro-2-fluorobenzeneboronic acid, (4-Chloro-2-fluorophenyl)metaboric acid, Dihydroxy(4-chloro-2-fluorophenyl)borane

AS32277G5
ACTIAS32277G5EA 0 CHF
ACTIAS32277G5
4-Chlor-2-fluorphenylboronsäure
4-Chlor-2-fluorphenylboronsäure
Formel: C₆H₅BClFO₂
MW: 174,37 g/mol
MDL Number: MFCD02684293
CAS-Nummer: 160591-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR