1-Boc-4-bromindol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-Boc-4-Bromoindole, 4-Bromo-1H-indole, N-BOC protected

AS30097G1 AS30097G5
ACTIAS30097G1EA 0 CHF
ACTIAS30097G1 ACTIAS30097G5
1-Boc-4-bromindol
1-Boc-4-bromindol
Formel: C₁₃H₁₄BrNO₂
MW: 296.16 g/mol
MDL Number: MFCD04117874
CAS-Nummer: 676448-17-2

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR