2,4-Difluor-5-nitrobenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

AS25245G100 AS25245G25 AS25245G5
ACTIAS25245G100EA 306 CHF
ACTIAS25245G100 ACTIAS25245G25 ACTIAS25245G5
2,4-Difluor-5-nitrobenzoesäure
2,4-Difluoro-5-nitrobenzoic acid
Formel: C₇H₃F₂NO₄
MW: 203.1 g/mol
MDL Number: MFCD04115713
CAS-Nummer: 153775-33-8
UN: 2811
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR