2,6-Dichlorphenethylamin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-(2-Aminoethyl)-1,3-dichlorobenzene, 1-Amino-2-(2,6-dichlorophenyl)ethane, 2-(2,6-Dichlorophenyl)ethylamine, 2,6-Dichlorophenethylamine

AS24939G100 AS24939G25 AS24939G5
ACTIAS24939G100EA 0 CHF
ACTIAS24939G100 ACTIAS24939G25 ACTIAS24939G5
2,6-Dichlorphenethylamin
2,6-Dichlorphenethylamin
Formel: C₈H₉Cl₂N
MW: 190.07 g/mol
MDL Number: MFCD00060612
CAS-Nummer: 14573-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR