2-(2-Aminoethoxy)benzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS22875G1 AS22875G5
ACTIAS22875G1EA 208 CHF
ACTIAS22875G1 ACTIAS22875G5
2-(2-Aminoethoxy)benzonitril
2-(2-Aminoethoxy)benzonitril
Formel: C₉H₁₀N₂O
MDL Number: MFCD16495889
CAS-Nummer: 57276-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR