2-Methyl-5-nitrophenylboronsäurepinakolester

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Methyl-5-nitrophenylboronic acid, pinacol ester, 2-Methyl-5-nitrophenylboronic Acid Pinacol Ester

AS22666G5
ACTIAS22666G5EA 0 CHF
ACTIAS22666G5
2-Methyl-5-nitrophenylboronsäurepinakolester
2-Methyl-5-nitrophenylboronsäurepinakolester
Formel: C₁₃H₁₈BNO₄
MW: 263.1 g/mol
MDL Number: MFCD09027081
CAS-Nummer: 957062-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR