2-Fluor-3-hydroxybenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

AS21934G1 AS21934G5
ACTIAS21934G1EA 0 CHF
ACTIAS21934G1 ACTIAS21934G5
2-Fluor-3-hydroxybenzoesäure
2-Fluoro-3-hydroxybenzoic acid
Formel: C₇H₅FO₃
MDL Number: MFCD01862066
CAS-Nummer: 91658-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR