4-Nitro-2-picolin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Methyl-4-nitropyridine

AS21655G25 AS21655G5
ACTIAS21655G25EA 0 CHF
ACTIAS21655G25 ACTIAS21655G5
4-Nitro-2-picolin
4-Nitro-2-picolin
Formel: C₆H₆N₂O₂
MDL Number: MFCD03095077
CAS-Nummer: 13508-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR