2,5-Dichlorbenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS21515G100 AS21515G25
ACTIAS21515G100EA 0 CHF
ACTIAS21515G100 ACTIAS21515G25
2,5-Dichlorbenzaldehyd
2,5-Dichlorobenzaldehyde
Formel: C₇H₄Cl₂O
MDL Number: MFCD00156140
CAS-Nummer: 6361-23-5
EINECS: 228-832-1

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR