4-Brom-2-nitroanilin

Lieferant: ACTIVATE SCIENTIFIC

AS20522G100
ACTIAS20522G100EA 0 CHF
ACTIAS20522G100
4-Brom-2-nitroanilin
4-Brom-2-nitroanilin
Formel: C₆H₅BrN₂O₂
MW: 217.02 g/mol
Siedepunkt: 309
Melting Pt: 110…113 °C
Dichte: 1.81 g/cm³ (20 °C)
MDL Number: MFCD00041312
CAS-Nummer: 875-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR