1-(2-Methoxyethyl)piperazin

Lieferant: ACTIVATE SCIENTIFIC

AS20286G25 AS20286G5
ACTIAS20286G25EA 0 CHF
ACTIAS20286G25 ACTIAS20286G5
1-(2-Methoxyethyl)piperazin
1-(2-Methoxyethyl)piperazin
Formel: C₇H₁₆N₂O
MW: 144.22 g/mol
MDL Number: MFCD00191214
CAS-Nummer: 13484-40-7
UN: 3267
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR