(S)-(+)-1-Boc-3-hydroxypyrrolidin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: tert-butyl (3S)-3-hydroxypyrrolidine-1-carboxylate, (S)-tert-Butyl 3-hydroxypyrrolidine-1-carboxylate

AS20192G100 AS20192G25
ACTIAS20192G100EA 0 CHF
ACTIAS20192G100 ACTIAS20192G25
(S)-(+)-1-Boc-3-hydroxypyrrolidin
(S)-(+)-1-Boc-3-hydroxypyrrolidin
Formel: C₉H₁₇NO₃
MDL Number: MFCD01317839
CAS-Nummer: 101469-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR