1-Boc-4-hydroxypiperidin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: N-Boc-4-piperidinol, tert-butyl 4-hydroxypiperidine-1-carboxylate, N-BOC-4-Hydroxypiperidine, tert-Butyl 4-hydroxypiperidine-1-carboxylate

AS20187G100 AS20187G25
ACTIAS20187G100EA 0 CHF
ACTIAS20187G100 ACTIAS20187G25
1-Boc-4-hydroxypiperidin
1-Boc-4-hydroxypiperidin
Formel: C₁₀H₁₉NO₃
MW: 201.27 g/mol
MDL Number: MFCD01075174
CAS-Nummer: 109384-19-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR