2-Aminonicotinsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Aminopyridine-3-carboxylic acid, 2-Amino-3-pyridinecarboxylic acid

AS20032G100 AS20032G25
ACTIAS20032G100EA 0 CHF
ACTIAS20032G100 ACTIAS20032G25
2-Aminonicotinsäure
2-Aminonicotinsäure
Formel: C₆H₆N₂O₂
Melting Pt: 297 °C
Dichte: 1.545 g/cm³ (20 °C)
MDL Number: MFCD00006318
CAS-Nummer: 5345-47-1
EINECS: 226-296-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR